4-(2-Bromophenyl)thiazole-2-carboxylic acid

Lieferant: Apollo Scientific

OR470186-1G
APOSOR470186-1GEA 601 EUR
APOSOR470186-1G
4-(2-Bromophenyl)thiazole-2-carboxylic acid
4-(2-Bromophenyl)thiazole-2-carboxylic acid
Formel: C₁₀H₆BrNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD07377577
CAS-Nummer: 1261268-93-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR