2-Amino-5-(2-bromophenyl)furan-3-carbonitrile

Lieferant: Apollo Scientific

OR470187-5G
APOSOR470187-5GEA 804 EUR
APOSOR470187-5G
2-Amino-5-(2-bromophenyl)furan-3-carbonitrile
2-Amino-5-(2-bromophenyl)furan-3-carbonitrile
Formel: C₁₁H₇BrN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD18375283
CAS-Nummer: 1261268-90-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR