2-Amino-5-(3-bromophenyl)furan-3-carbonitrile

Lieferant: Apollo Scientific

OR470188-5G
APOSOR470188-5GEA 804 EUR
APOSOR470188-5G
2-Amino-5-(3-bromophenyl)furan-3-carbonitrile
2-Amino-5-(3-bromophenyl)furan-3-carbonitrile
Formel: C₁₁H₇BrN₂O
Molecular Weight: 263,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18375284
CAS-Nummer: 1261268-89-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR