Methyl 2-(5-Amino-4-cyano-2-furyl)benzoate

Lieferant: Apollo Scientific

OR470190-5G
APOSOR470190-5GEA 1010 EUR
APOSOR470190-5G
Methyl 2-(5-Amino-4-cyano-2-furyl)benzoate
Methyl 2-(5-Amino-4-cyano-2-furyl)benzoate
Formel: C₁₃H₁₀N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18375285
CAS-Nummer: 1261268-76-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR