Methyl 3-(5-Amino-4-cyano-2-furyl)benzoate

Lieferant: Apollo Scientific

OR470191-5G
APOSOR470191-5GEA 916 EUR
APOSOR470191-5G
Methyl 3-(5-Amino-4-cyano-2-furyl)benzoate
Methyl 3-(5-Amino-4-cyano-2-furyl)benzoate
Formel: C₁₃H₁₀N₂O₃
Molecular Weight: 242,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18375286
CAS-Nummer: 1261269-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR