Methyl 4-(5-Amino-4-cyano-2-furyl)benzoate

Lieferant: Apollo Scientific

OR470192-5G
APOSOR470192-5GEA 916 EUR
APOSOR470192-5G
Methyl 4-(5-Amino-4-cyano-2-furyl)benzoate
Methyl 4-(5-Amino-4-cyano-2-furyl)benzoate
Formel: C₁₃H₁₀N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18375287
CAS-Nummer: 1261268-86-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR