2-(2-Nitrophenyl)imidazole

Lieferant: Apollo Scientific

OR470196-1G
APOSOR470196-1GEA 708 EUR
APOSOR470196-1G
2-(2-Nitrophenyl)imidazole
2-(2-Nitrophenyl)imidazole
Formel: C₉H₇N₃O₂
Molecular Weight: 189,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863372
CAS-Nummer: 4205-06-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR