Methyl 1-Benzyl-4-hydroxypiperidine-3-carboxylate
Lieferant: Apollo Scientific
OR470204-1G
APOSOR470204-1GEA
601
EUR
APOSOR470204-1G
Methyl 1-Benzyl-4-hydroxypiperidine-3-carboxylate
Methyl 1-Benzyl-4-hydroxypiperidine-3-carboxylate
Formel:
C₁₄H₁₉NO₃ Molecular Weight: 249,306 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18071560 CAS-Nummer: 955998-64-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...