2-[Bis(methylsulphonyl)amino]benzonitrile

Lieferant: Apollo Scientific

OR470206-25G OR470206-5G
APOSOR470206-25GEA 1300 EUR
APOSOR470206-25G APOSOR470206-5G
2-[Bis(methylsulphonyl)amino]benzonitrile
2-[Bis(methylsulphonyl)amino]benzonitrile
Formel: C₉H₁₀N₂O₄S₂
Molecular Weight: 274.317 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01648439
CAS-Nummer: 235100-91-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR