6-Ethoxy-1,3-dimethyluracil

Lieferant: Apollo Scientific

OR470208-5G
APOSOR470208-5GEA 1330 EUR
APOSOR470208-5G
6-Ethoxy-1,3-dimethyluracil
6-Ethoxy-1,3-dimethyluracil
Formel: C₈H₁₂N₂O₃
Molecular Weight: 184,192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11656357
CAS-Nummer: 93787-99-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR