6-Nitrospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-4-one

Lieferant: Apollo Scientific

OR470220-5G
APOSOR470220-5GEA 1120 EUR
APOSOR470220-5G
6-Nitrospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-4-one
6-Nitrospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-4-one
Formel: C₁₂H₁₃N₃O₃
Molecular Weight: 247.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651722
CAS-Nummer: 158439-85-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR