4-Oxospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclobutane]-5-carbonitrile

Lieferant: Apollo Scientific

OR470221-5G
APOSOR470221-5GEA 1220 EUR
APOSOR470221-5G
4-Oxospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclobutane]-5-carbonitrile
4-Oxospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclobutane]-5-carbonitrile
Formel: C₁₂H₁₁N₃O
Molecular Weight: 213.235 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651724
CAS-Nummer: 1272756-09-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR