Ethyl 4-Oxospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclobutane]-5-carboxylate
Lieferant: Apollo Scientific
OR470222-5G
APOSOR470222-5GEA
1410
EUR
APOSOR470222-5G
Ethyl 4-Oxospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclobutane]-5-carboxylate
Ethyl 4-Oxospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclobutane]-5-carboxylate
Formel:
C₁₄H₁₆N₂O₃ Molecular Weight: 260,288 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18651725 CAS-Nummer: 1272756-15-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...