4-Oxospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclobutane]-5-carboxylic acid

Lieferant: Apollo Scientific

OR470223-5G
APOSOR470223-5GEA 1410 EUR
APOSOR470223-5G
4-Oxospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclobutane]-5-carboxylic acid
4-Oxospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclobutane]-5-carboxylic acid
Formel: C₁₂H₁₂N₂O₃
Molecular Weight: 232,235 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651726
CAS-Nummer: 1272756-04-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR