6-Bromospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclobutane]-4-one
Lieferant: Apollo Scientific
OR470224-5G
APOSOR470224-5GEA
1220
EUR
APOSOR470224-5G
6-Bromospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclobutane]-4-one
6-Bromospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclobutane]-4-one
Formel:
C₁₁H₁₁BrN₂O Molecular Weight: 267,122 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18651728 CAS-Nummer: 1272756-13-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...