Ethyl 4-Oxospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-5-carboxylate

Lieferant: Apollo Scientific

OR470225-5G
APOSOR470225-5GEA 1410 EUR
APOSOR470225-5G
Ethyl 4-Oxospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-5-carboxylate
Ethyl 4-Oxospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-5-carboxylate
Formel: C₁₅H₁₈N₂O₃
Molecular Weight: 274,315 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651730
CAS-Nummer: 1272756-22-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR