4-Oxospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-5-carboxylic acid

Lieferant: Apollo Scientific

OR470226-5G
APOSOR470226-5GEA 1410 EUR
APOSOR470226-5G
4-Oxospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-5-carboxylic acid
4-Oxospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-5-carboxylic acid
Formel: C₁₃H₁₄N₂O₃
Molecular Weight: 246,262 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651731
CAS-Nummer: 1272756-14-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR