6-Bromospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-4-one

Lieferant: Apollo Scientific

OR470227-5G
APOSOR470227-5GEA 1120 EUR
APOSOR470227-5G
6-Bromospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-4-one
6-Bromospiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclopentane]-4-one
Formel: C₁₂H₁₃BrN₂O
Molecular Weight: 281,148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651733
CAS-Nummer: 1272756-17-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR