2-(3-Oxo-1,4-diazaspiro[4.4]non-2-yl)benzonitrile
Lieferant: Apollo Scientific
OR470228-5G
APOSOR470228-5GEA
1120
EUR
APOSOR470228-5G
2-(3-Oxo-1,4-diazaspiro[4.4]non-2-yl)benzonitrile
2-(3-Oxo-1,4-diazaspiro[4.4]non-2-yl)benzonitrile
Formel:
C₁₄H₁₅N₃O Molecular Weight: 241,288 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18651735 CAS-Nummer: 1272755-94-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...