2-(3-Oxo-1,4-diazaspiro[4.4]non-2-yl)benzoic acid

Lieferant: Apollo Scientific

OR470229-5G
APOSOR470229-5GEA 1120 EUR
APOSOR470229-5G
2-(3-Oxo-1,4-diazaspiro[4.4]non-2-yl)benzoic acid
2-(3-Oxo-1,4-diazaspiro[4.4]non-2-yl)benzoic acid
Formel: C₁₄H₁₆N₂O₃
Molecular Weight: 260,288 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651737
CAS-Nummer: 1272755-92-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR