3-(3-Bromophenyl)-1,4-diazaspiro[4.4]nonan-2-one

Lieferant: Apollo Scientific

OR470230-5G
APOSOR470230-5GEA 1120 EUR
APOSOR470230-5G
3-(3-Bromophenyl)-1,4-diazaspiro[4.4]nonan-2-one
3-(3-Bromophenyl)-1,4-diazaspiro[4.4]nonan-2-one
Formel: C₁₃H₁₅BrN₂O
Molecular Weight: 295,175 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651738
CAS-Nummer: 1272755-85-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR