3-(3-Nitrophenyl)-1,4-diazaspiro[4.4]nonan-2-one

Lieferant: Apollo Scientific

OR470231-5G
APOSOR470231-5GEA 1120 EUR
APOSOR470231-5G
3-(3-Nitrophenyl)-1,4-diazaspiro[4.4]nonan-2-one
3-(3-Nitrophenyl)-1,4-diazaspiro[4.4]nonan-2-one
Formel: C₁₃H₁₅N₃O₃
Molecular Weight: 261,277 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651739
CAS-Nummer: 1272755-82-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR