4-(3-Oxo-1,4-diazaspiro[4.4]non-2-yl)benzonitrile

Lieferant: Apollo Scientific

OR470232-5G
APOSOR470232-5GEA 1120 EUR
APOSOR470232-5G
4-(3-Oxo-1,4-diazaspiro[4.4]non-2-yl)benzonitrile
4-(3-Oxo-1,4-diazaspiro[4.4]non-2-yl)benzonitrile
Formel: C₁₄H₁₅N₃O
Molecular Weight: 241,288 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651740
CAS-Nummer: 1272755-80-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR