3-(2-Bromophenyl)-1,4-diazaspiro[4.4]nonan-2-one
Lieferant: Apollo Scientific
OR470233-5G
APOSOR470233-5GEA
1120
EUR
APOSOR470233-5G
3-(2-Bromophenyl)-1,4-diazaspiro[4.4]nonan-2-one
3-(2-Bromophenyl)-1,4-diazaspiro[4.4]nonan-2-one
Formel:
C₁₃H₁₅BrN₂O Molecular Weight: 295,175 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18651741 CAS-Nummer: 1272755-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...