3-[4-(Methylsulphonyl)phenyl]-1,4-diazaspiro[4.4]nonan-2-one

Lieferant: Apollo Scientific

OR470234-5G
APOSOR470234-5GEA 1120 EUR
APOSOR470234-5G
3-[4-(Methylsulphonyl)phenyl]-1,4-diazaspiro[4.4]nonan-2-one
3-[4-(Methylsulphonyl)phenyl]-1,4-diazaspiro[4.4]nonan-2-one
Formel: C₁₄H₁₈N₂O₃S
Molecular Weight: 294,369 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651743
CAS-Nummer: 1272755-86-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR