2-(7-Oxo-5,8-diazaspiro[3.4]oct-6-yl)benzonitrile

Lieferant: Apollo Scientific

OR470235-5G
APOSOR470235-5GEA 1220 EUR
APOSOR470235-5G
2-(7-Oxo-5,8-diazaspiro[3.4]oct-6-yl)benzonitrile
2-(7-Oxo-5,8-diazaspiro[3.4]oct-6-yl)benzonitrile
Formel: C₁₃H₁₃N₃O
Molecular Weight: 227,262 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651745
CAS-Nummer: 1272755-93-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR