2-(7-Oxo-5,8-diazaspiro[3.4]oct-6-yl)benzoic acid

Lieferant: Apollo Scientific

OR470236-5G
APOSOR470236-5GEA 1220 EUR
APOSOR470236-5G
2-(7-Oxo-5,8-diazaspiro[3.4]oct-6-yl)benzoic acid
2-(7-Oxo-5,8-diazaspiro[3.4]oct-6-yl)benzoic acid
Formel: C₁₃H₁₄N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18651747
CAS-Nummer: 1272755-83-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR