7-(3-Nitrophenyl)-5,8-diazaspiro[3.4]octan-6-one

Lieferant: Apollo Scientific

OR470238-5G
APOSOR470238-5GEA 1220 EUR
APOSOR470238-5G
7-(3-Nitrophenyl)-5,8-diazaspiro[3.4]octan-6-one
7-(3-Nitrophenyl)-5,8-diazaspiro[3.4]octan-6-one
Formel: C₁₂H₁₃N₃O₃
Molecular Weight: 247,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651749
CAS-Nummer: 1272755-97-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR