7-(2-Bromophenyl)-5,8-diazaspiro[3.4]octan-6-one

Lieferant: Apollo Scientific

OR470240-5G
APOSOR470240-5GEA 1220 EUR
APOSOR470240-5G
7-(2-Bromophenyl)-5,8-diazaspiro[3.4]octan-6-one
7-(2-Bromophenyl)-5,8-diazaspiro[3.4]octan-6-one
Formel: C₁₂H₁₃BrN₂O
Molecular Weight: 281,148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18651751
CAS-Nummer: 1272755-95-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR