7-[4-(Methylsulphonyl)phenyl]-5,8-diazaspiro[3.4]octan-6-one

Lieferant: Apollo Scientific

OR470241-5G
APOSOR470241-5GEA 1220 EUR
APOSOR470241-5G
7-[4-(Methylsulphonyl)phenyl]-5,8-diazaspiro[3.4]octan-6-one
7-[4-(Methylsulphonyl)phenyl]-5,8-diazaspiro[3.4]octan-6-one
Formel: C₁₃H₁₆N₂O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD18651753
CAS-Nummer: 1272755-84-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR