6-Chloro-5-methyl-1H-indazole
Lieferant: Apollo Scientific
OR470242-1G
APOSOR470242-1GEA
125
EUR
APOSOR470242-1G
6-Chloro-5-methyl-1H-indazole
6-Chloro-5-methyl-1H-indazole
Formel:
C₈H₇ClN₂ Molecular Weight: 166,608 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09880179 CAS-Nummer: 1000341-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...