Methyl 4-(1,1-Dioxothiomorpholino)butyrate

Lieferant: Apollo Scientific

OR470243-1G
APOSOR470243-1GEA 713 EUR
APOSOR470243-1G
Methyl 4-(1,1-Dioxothiomorpholino)butyrate
Methyl 4-(1,1-Dioxothiomorpholino)butyrate
Formel: C₉H₁₇NO₄S
Molecular Weight: 235,301 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09880001
CAS-Nummer: 1000342-02-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR