1-Acetylindole-2-carboxylic acid

Lieferant: Apollo Scientific

Synonyme: 1-Acetyl-1H-indole-2-carboxylic acid

OR470244-5G
APOSOR470244-5GEA 1410 EUR
APOSOR470244-5G
1-Acetylindole-2-carboxylic acid
1-Acetylindole-2-carboxylic acid
Formel: C₁₁H₉NO₃
Molecular Weight: 203,194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02258917
CAS-Nummer: 10441-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR