4-Methyl-1-piperazinepropionic acid hydrochloride

Lieferant: Apollo Scientific

OR470248-5G
APOSOR470248-5GEA 322 EUR
APOSOR470248-5G
4-Methyl-1-piperazinepropionic acid hydrochloride
4-Methyl-1-piperazinepropionic acid hydrochloride
Formel: C₈H₁₆N₂O₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD03452741
CAS-Nummer: 775349-40-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR