1-Boc-4-hydroxy-4-(nitromethyl)piperidine

Lieferant: Apollo Scientific

OR470249-5G
APOSOR470249-5GEA 1350 EUR
APOSOR470249-5G
1-Boc-4-hydroxy-4-(nitromethyl)piperidine
1-Boc-4-hydroxy-4-(nitromethyl)piperidine
Formel: C₁₁H₂₀N₂O₅
Molecular Weight: 260,287 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12963548
CAS-Nummer: 819800-93-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR