3-Iodo-1-methyl-4-nitro-1H-pyrrolo[2,3-b]pyridine

Lieferant: Apollo Scientific

OR470252-1G
APOSOR470252-1GEA 1540 EUR
APOSOR470252-1G
3-Iodo-1-methyl-4-nitro-1H-pyrrolo[2,3-b]pyridine
3-Iodo-1-methyl-4-nitro-1H-pyrrolo[2,3-b]pyridine
Formel: C₈H₆IN₃O₂
Molecular Weight: 303,057 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423101
CAS-Nummer: 1398534-56-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR