4-Benzyloxy-3-nitrobenzoic acid

Lieferant: Apollo Scientific

OR470253-5G
APOSOR470253-5GEA 601 EUR
APOSOR470253-5G
4-Benzyloxy-3-nitrobenzoic acid
4-Benzyloxy-3-nitrobenzoic acid
Formel: C₁₄H₁₁NO₅
Molecular Weight: 273,241 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03425435
CAS-Nummer: 17903-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR