3-Chloro-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyridine

Lieferant: Apollo Scientific

OR470254-1G
APOSOR470254-1GEA 1210 EUR
APOSOR470254-1G
3-Chloro-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyridine
3-Chloro-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyridine
Formel: C₆H₈ClN₃
Molecular Weight: 157,601 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22423103
CAS-Nummer: 1398534-61-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR