1-Methyl-6-nitrobenzimidazole

Lieferant: Apollo Scientific

OR470255-5G
APOSOR470255-5GEA 1210 EUR
APOSOR470255-5G
1-Methyl-6-nitrobenzimidazole
1-Methyl-6-nitrobenzimidazole
Formel: C₈H₇N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00962814
CAS-Nummer: 5381-79-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR