3-(3-Indolyl)-2-methylpropionic acid

Lieferant: Apollo Scientific

OR470258-5G
APOSOR470258-5GEA 1410 EUR
APOSOR470258-5G
3-(3-Indolyl)-2-methylpropionic acid
3-(3-Indolyl)-2-methylpropionic acid
Formel: C₁₂H₁₃NO₂
Molecular Weight: 203,237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01860840
CAS-Nummer: 15142-91-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR