2-(Phenethoxymethyl)pyrrolidine

Lieferant: Apollo Scientific

OR470261-1G
APOSOR470261-1GEA 713 EUR
APOSOR470261-1G
2-(Phenethoxymethyl)pyrrolidine
2-(Phenethoxymethyl)pyrrolidine
Formel: C₁₃H₁₉NO
Lagertemperatur: Raumtemperatur
MDL: MFCD06248668
CAS-Nummer: 883546-84-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR