Methyl 2-Amino-2-(3,5-dichlorophenyl)acetate hydrochloride

Lieferant: Apollo Scientific

OR470264-5G
APOSOR470264-5GEA 1540 EUR
APOSOR470264-5G
Methyl 2-Amino-2-(3,5-dichlorophenyl)acetate hydrochloride
Methyl 2-Amino-2-(3,5-dichlorophenyl)acetate hydrochloride
Formel: C₉H₉Cl₂NO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD04115513
CAS-Nummer: 1137447-08-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR