2-Amino-2-(3-chlorophenyl)acetic acid hydrochloride

Lieferant: Apollo Scientific

OR470265-5G
APOSOR470265-5GEA 916 EUR
APOSOR470265-5G
2-Amino-2-(3-chlorophenyl)acetic acid hydrochloride
2-Amino-2-(3-chlorophenyl)acetic acid hydrochloride
Formel: C₈H₈ClNO₂·ClH
Molecular Weight: 222,069 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410535
CAS-Nummer: 1214196-70-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR