3-Amino-3-cyclopentylpropanoic acid

Lieferant: Apollo Scientific

OR470266-5G
APOSOR470266-5GEA 1540 EUR
APOSOR470266-5G
3-Amino-3-cyclopentylpropanoic acid
3-Amino-3-cyclopentylpropanoic acid
Formel: C₈H₁₅NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03002509
CAS-Nummer: 682804-23-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR