Ethyl 3-(6-Bromo-3-pyridinyl)-3-oxopropanoate

Lieferant: Apollo Scientific

OR470268-5G
APOSOR470268-5GEA 1120 EUR
APOSOR470268-5G
Ethyl 3-(6-Bromo-3-pyridinyl)-3-oxopropanoate
Ethyl 3-(6-Bromo-3-pyridinyl)-3-oxopropanoate
Formel: C₁₀H₁₀BrNO₃
Molecular Weight: 272,095 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07370057
CAS-Nummer: 916791-37-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR