6-(3-Chlorophenyl)nicotinic acid

Lieferant: Apollo Scientific

OR470269-5G
APOSOR470269-5GEA 1540 EUR
APOSOR470269-5G
6-(3-Chlorophenyl)nicotinic acid
6-(3-Chlorophenyl)nicotinic acid
Formel: C₁₂H₈ClNO₂
Molecular Weight: 233.65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410393
CAS-Nummer: 887976-19-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR