2-(Boc-amino)-2-(2-naphthyl)acetic acid

Lieferant: Apollo Scientific

Synonyme: 2-((tert-Butoxycarbonyl)amino)-2-(naphthalen-2-yl)acetic acid

OR470271-5G
APOSOR470271-5GEA 1330 EUR
APOSOR470271-5G
2-(Boc-amino)-2-(2-naphthyl)acetic acid
2-(Boc-amino)-2-(2-naphthyl)acetic acid
Formel: C₁₇H₁₉NO₄
Molecular Weight: 301,337 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02683164
CAS-Nummer: 33741-79-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR