2-Amino-2-[4-(2-pyridyl)phenyl]acetic acid hydrochloride

Lieferant: Apollo Scientific

OR470272-5G
APOSOR470272-5GEA 1620 EUR
APOSOR470272-5G
2-Amino-2-[4-(2-pyridyl)phenyl]acetic acid hydrochloride
2-Amino-2-[4-(2-pyridyl)phenyl]acetic acid hydrochloride
Formel: C₁₃H₁₂N₂O₂·ClH
Molecular Weight: 264,708 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07371460
CAS-Nummer: 1135818-89-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR