Ethyl trans-3-Oxo-3-(2-phenylcyclopropyl)propanoate
Lieferant: Apollo Scientific
OR470273-5G
APOSOR470273-5GEA
1330
EUR
APOSOR470273-5G
Ethyl trans-3-Oxo-3-(2-phenylcyclopropyl)propanoate
Ethyl trans-3-Oxo-3-(2-phenylcyclopropyl)propanoate
Formel:
C₁₄H₁₆O₃ Molecular Weight: 232,275 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09025936 CAS-Nummer: 324570-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...