6-(3-Nitrophenyl)picolinsäure

Lieferant: Apollo Scientific

OR470274-5G
APOSOR470274-5GEA 1540 EUR
APOSOR470274-5G
6-(3-Nitrophenyl)picolinsäure
6-(3-Nitrophenyl)picolinsäure
Formel: C₁₂H₈N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD06410075
CAS-Nummer: 80021-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR